Invitrogen
Manufacturer Code:PA534541
Catalog # PIPA534541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the carboxy terminus of human GBP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GBP-5; GBP-TA antigen; GBP5 Guanylate binding protein family member 5 GBP-5 GTP-binding protein 5 Guanine nucleotide-binding protein 5 GBP-TA antigen; GTP-binding protein 5; Guanine nucleotide-binding protein 5; guanylate nucleotide binding protein 5; Guanylate-binding protein 5; muGBP-5
Gene Aliases: 5330409J06Rik; GBP-5; GBP5; Gbp5a; UNQ2427/PRO4987
UniProt ID: (Human) Q96PP8, (Mouse) Q8CFB4
Entrez Gene ID: (Human) 115362, (Mouse) 229898, (Rat) 362050
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.