Proteintech
Manufacturer Code:118541AP150UL
Catalog # 50-559-541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GBP2 Fusion Protein Ag2425 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GBP 2 GBP2 GTP binding protein 2 HuGBP 2; GBP-2; GTP-binding protein 2; Guanine nucleotide-binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; Interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; p67
Gene Aliases: GBP2
UniProt ID: (Human) P32456, (Rat) Q63663
Entrez Gene ID: (Human) 2634, (Rat) 171164
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.