Invitrogen
Manufacturer Code:PA5116392
Catalog # PIPA5116392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 154-185 amino acids from the Central region of human GBGT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3GALNT Fgs Forssman glycolipid synthase forssman glycolipid synthase-like protein forssman glycolipid synthetase Forssman glycolipid synthetase (FS) FS GBGT1 globoside alpha-13-N-acetylgalactosaminyltransferase 1 LOC499764 similar to globoside alpha-13-N-acetylgalactosaminyltransferase 1 UNQ2513 UNQ2513/PRO6002; Forssman glycolipid synthase-like protein; Forssman glycolipid synthetase (FS); Globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases: A3GALNT; FS; GBGT1; UNQ2513; UNQ2513/PRO6002
UniProt ID: (Human) Q8N5D6
Entrez Gene ID: (Human) 26301
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.