Novus Biologicals
Manufacturer Code:NBP214040
Catalog # NBP214040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: HMAILADKANGIMAAWREESHLNRHFISNKPSKVLSPEYLWDDRKPQPPS LKLIRFSTLDKDISCLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A3GALNT EC 2.4.1.- Forssman glycolipid synthase-like protein Forssman glycolipid synthetase (FS) Forssman synthetase FSUNQ2513 globoside alpha-13-N-acetylgalactosaminyltransferase 1 MGC44848 UDP-GalNAc; Forssman glycolipid synthase-like protein; Forssman glycolipid synthetase (FS); Globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases: A3GALNT; FS; GBGT1; UNQ2513; UNQ2513/PRO6002
UniProt ID: (Human) Q8N5D6
Entrez Gene ID: (Human) 26301
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.