Novus Biologicals
Manufacturer Code:NBP106525
Catalog # NBP106525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human golgi-specific brefeldin A resistance factor 1 using the numbering given in entry NP_004184.1 (GeneID 8729). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARF1GEF BFA-resistant GEF 1 FLJ21263 golgi brefeldin A resistant guanine nucleotide exchange factor 1 golgi-specific brefeldin A resistance factor 1 Golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1 KIAA0248FLJ21500 MGC134877 MGC134878; BFA-resistant GEF 1; Golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1
Gene Aliases: ARF1GEF; GBF1; KIAA0248
UniProt ID: (Human) Q92538
Entrez Gene ID: (Human) 8729
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.