Proteintech
Manufacturer Code:203131AP150UL
Catalog # 50-173-2824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GBE1 Fusion Protein Ag14154 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-glucan-branching enzyme; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; Brancher enzyme; Brancher enzyme GBE GBE1 Glycogen branching enzyme; glycogen branching enzyme; Glycogen-branching enzyme
Gene Aliases: 2310045H19Rik; 2810426P10Rik; APBD; D16Ertd536e; GBE; GBE1; GSD4
UniProt ID: (Human) Q04446, (Mouse) Q9D6Y9
Entrez Gene ID: (Human) 2632, (Mouse) 74185, (Rat) 288333
Molecular Function:
amylase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.