Invitrogen
Manufacturer Code:MA526169
Catalog # PIMA526169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B8 |
Immunogen | Full length human recombit protein of GBAS produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; glioblastoma amplified sequence; Glioblastoma-amplified sequence; NipSnap2; NIPSNAP2 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2 protein NipSnap homolog 2; Protein NipSnap homolog 2
Gene Aliases: AV006093; GBAS; NIPSNAP2
UniProt ID: (Human) O75323
Entrez Gene ID: (Human) 2631, (Mouse) 14467, (Rat) 498174, (Dog) 479700, (Rhesus monkey) 715352
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.