Novus Biologicals
Manufacturer Code:NBP270804B
Catalog # NBP270804B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B8 |
Immunogen | Full length recombinant protein of human GBAS (NP_001474) produced in E.coli. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; glioblastoma amplified sequence Glioblastoma-amplified sequence NIPSNAP2NipSnap24-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2 protein NipSnap homolog 2; Glioblastoma-amplified sequence; NipSnap2; Protein NipSnap homolog 2
Gene Aliases: GBAS; NIPSNAP2
UniProt ID: (Human) O75323
Entrez Gene ID: (Human) 2631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.