Invitrogen
Manufacturer Code:PA534899
Catalog # PIPA534899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contains a sequence corresponding to a region within amino acids 77 and 265 (O75323) of GBAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; Glioblastoma-amplified sequence; NipSnap2; NIPSNAP2 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2 protein NipSnap homolog 2; Protein NipSnap homolog 2
Gene Aliases: GBAS; NIPSNAP2
UniProt ID: (Human) O75323
Entrez Gene ID: (Human) 2631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.