OriGene
Manufacturer Code:TA700134
Catalog # 50-167-7667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F10 |
Immunogen | Full length human recombit protein of human HPGD produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CBG CBGL1 GLUC KLRP; Cytosolic beta-glucosidase; Cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Glucosidase beta acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; Klotho-related protein; KLrP
Gene Aliases: CBG; CBGL1; GBA3; GLUC; KLRP
UniProt ID: (Human) Q9H227
Entrez Gene ID: (Human) 57733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.