Invitrogen
Manufacturer Code:MA525607
Catalog # PIMA525607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full length human recombit protein of GATM produced in HEK293 cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: glycine amidinotransferase (L-arginine:glycine amidinotransferase); Glycine amidinotransferase mitochondrial L-arginine:glycine amidinotransferaseTransamidinase GATM AGAT; Glycine amidinotransferase, mitochondrial; L-arginine:glycine amidinotransferase; testicular secretory protein Li 19; Transamidinase
Gene Aliases: AGAT; AT; CCDS3; GATM
UniProt ID: (Human) P50440
Entrez Gene ID: (Human) 2628
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.