Invitrogen
Manufacturer Code:PA587228
Catalog # PIPA587228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GATM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AT; glycine amidinotransferase (L-arginine:glycine amidinotransferase); Glycine amidinotransferase mitochondrial L-arginine:glycine amidinotransferaseTransamidinase GATM AGAT; Glycine amidinotransferase, mitochondrial; L-arginine: glycine amidinotransferase; L-arginine:glycine amidinotransferase; testicular secretory protein Li 19; Transamidinase
Gene Aliases: 1810003P21Rik; AGAT; AI314789; AT; CCDS3; GATM
UniProt ID: (Human) P50440, (Mouse) Q9D964, (Rat) P50442
Entrez Gene ID: (Human) 2628, (Mouse) 67092, (Rat) 81660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.