Invitrogen
Manufacturer Code:PA5109756
Catalog # PIPA5109756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 124-423 of human GATM (NP_001473.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810003P21Rik AGAT AI314789 AT CCDS3 GATM Glycine amidinotransferase mitochondrial L-arginine:glycine amidinotransferase Transamidinase; AT; glycine amidinotransferase (L-arginine:glycine amidinotransferase); Glycine amidinotransferase, mitochondrial; L-arginine: glycine amidinotransferase; L-arginine:glycine amidinotransferase; testicular secretory protein Li 19; Transamidinase
Gene Aliases: 1810003P21Rik; AGAT; AI314789; AT; CCDS3; GATM
UniProt ID: (Human) P50440, (Mouse) Q9D964, (Rat) P50442
Entrez Gene ID: (Human) 2628, (Mouse) 67092, (Rat) 81660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.