Invitrogen
Manufacturer Code:PA576426
Catalog # PIPA576426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human GATC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15E1.2; gatC-like protein; Glu-AdT subunit C; glu-AdT subunit C {ECO:0000255|HAMAP-Rule:MF_03149}; Glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03149}; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog
Gene Aliases: 2010003O18Rik; Gatc; RGD1309698
UniProt ID: (Rat) D3ZY68, (Mouse) Q8CBY0
Entrez Gene ID: (Rat) 360821, (Mouse) 384281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.