Invitrogen
Manufacturer Code:PA5117145
Catalog # PIPA5117145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human GATC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15E1.2 2010003O18Rik Gatc gatC-like protein glu-AdT subunit C glu-AdT subunit C {ECO:0000255 glutamyl-tRNA amidotransferase subunit C glutamyl-tRNA(Gln) amidotransferase subunit C mitochondrial glutamyl-tRNA(Gln) amidotransferase subunit C mitochondrial {ECO:0000255 glutamyl-tRNA(Gln) amidotransferase subunit C glutamyl-tRNA(Gln) amidotransferase subunit C homolog HAMAP-Rule:MF_03149} Protein 15E1.2 RGD1309698; GATC; gatC-like protein; Glu-AdT subunit C; glu-AdT subunit C {ECO:0000255|HAMAP-Rule:MF_03149}; Glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03149}; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog; Protein 15E1.2
Gene Aliases: 15E1.2; 2010003O18Rik; GATC; RGD1309698
UniProt ID: (Human) O43716, (Rat) D3ZY68, (Mouse) Q8CBY0
Entrez Gene ID: (Human) 283459, (Rat) 360821, (Mouse) 384281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.