Proteintech
Manufacturer Code:231831AP150UL
Catalog # 50-173-4014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GATC Fusion Protein Ag19713 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15E1.2 GATC GatC like protein Protein 15E1.2; GATC; gatC-like protein; Glu-AdT subunit C; Glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog; Protein 15E1.2
Gene Aliases: 15E1.2; 2010003O18Rik; GATC
UniProt ID: (Human) O43716, (Mouse) Q8CBY0
Entrez Gene ID: (Human) 283459, (Mouse) 384281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.