Proteintech
Manufacturer Code:122941AP150UL
Catalog # 50-172-7867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GATAD2A Fusion Protein Ag2947 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATA zinc finger domain-containing protein 2A; GATAD2A Hp66alpha p66 alpha; Hp66alpha; p66 alpha; Transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha
Gene Aliases: 1110066C11Rik; BC031407; C80248; GATAD2A; p66alpha; RGD1310596
UniProt ID: (Human) Q86YP4, (Mouse) Q8CHY6
Entrez Gene ID: (Human) 54815, (Mouse) 234366, (Rat) 290669
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.