Invitrogen
Manufacturer Code:PA141241
Catalog # 01-675-595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 137-150 of mouse p66a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110066C11Rik BC031407 C80248 MGC29315 MGC36975; GATA zinc finger domain-containing protein 2A; Hp66alpha; p66 alpha; Transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha
Gene Aliases: 1110066C11Rik; BC031407; C80248; GATAD2A; p66alpha; RGD1310596
UniProt ID: (Human) Q86YP4, (Mouse) Q8CHY6
Entrez Gene ID: (Human) 54815, (Mouse) 234366, (Rat) 290669
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.