Invitrogen
Manufacturer Code:PA540559
Catalog # PIPA540559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GATA6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding protein GATA-GT2; GATA-binding factor 6; GATA-binding protein 6; Transcription factor GATA-6; zinc-finger transcription factor
Gene Aliases: AA410133; GATA-6; GATA6
UniProt ID: (Human) Q92908, (Pig) Q95JA5, (Rat) P46153, (Mouse) Q61169
Entrez Gene ID: (Human) 2627, (Bovine) 654400, (Pig) 397600, (Dog) 490520, (Rat) 29300, (Mouse) 14465
Molecular Function: nuclease nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.