Invitrogen
Manufacturer Code:PA1104X
Catalog # PIPA1104X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence KYSGQDGLYIGVSLASPAE & KRFGAAGADASDSRAFPARE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATA-binding factor 6; GATA-binding factor 6 GATA-binding protein 6; GATA-binding protein 6; Transcription factor GATA-6
Gene Aliases: AA410133; GATA-6; GATA6
UniProt ID: (Human) Q92908, (Mouse) Q61169
Entrez Gene ID: (Human) 2627, (Mouse) 14465
Molecular Function: nuclease nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.