Invitrogen
Manufacturer Code:PA568946
Catalog # PIPA568946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse GATA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATA-binding factor 5; GATA-binding protein 5; GATA5 GATA binding protein 5 bB379O24.1 GATA binding factor-5 GATA-binding protein 5 transcription factor GATA-5; Transcription factor GATA-5; Transcription factor GATA-5 (GATA binding factor-5)
Gene Aliases: GATA5
UniProt ID: (Bovine) Q3SZJ5, (Mouse) P97489
Entrez Gene ID: (Dog) 485961, (Bovine) 504784, (Mouse) 14464, (Rat) 499951
Molecular Function: nuclease nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.