OriGene
Manufacturer Code:TA500091
Catalog # 50-167-1394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9F9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 296-442 of human GATA4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ASD2 TACHD TOF VSD1; GATA-binding factor 4; Transcription factor GATA-4
Gene Aliases: ASD2; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694
Entrez Gene ID: (Human) 2626
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.