Invitrogen
Manufacturer Code:MA515532
Catalog # PIMA515532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6H10 |
Immunogen | Purified recombit fragment of human GATA4 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding protein GATA-GT2; GATA-binding factor 4; GATA-binding protein 4; MGC126629 GATA4; Transcription factor GATA-4
Gene Aliases: ASD2; Gata-4; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694, (Mouse) Q08369, (Rat) P46152
Entrez Gene ID: (Human) 2626, (Mouse) 14463, (Rat) 54254
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.