Invitrogen
Manufacturer Code:PA1102X
Catalog # PIPA1102X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Pooled peptides: RKEGIQTRKRKPKNLNKSK & CKQDSWNSLVLADSHGDIITA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATA-binding factor 4; GATA-binding protein 4; GATA4 GATA binding factor 4; Transcription factor GATA-4
Gene Aliases: ASD2; Gata-4; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694, (Mouse) Q08369
Entrez Gene ID: (Human) 2626, (Mouse) 14463
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.