OriGene
Manufacturer Code:UM870110
Catalog # 50-168-5559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB218 |
Immunogen | Human recombit protein fragment corresponding to amino acids 155-443 of human GATA3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GATA-binding factor 3; HDR HDRS; Trans-acting T-cell-specific transcription factor GATA-3
Gene Aliases: GATA3; HDR; HDRS
UniProt ID: (Human) P23771
Entrez Gene ID: (Human) 2625
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.