Invitrogen
Manufacturer Code:PA1099X
Catalog # PIPA1099X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N-terminal truncated human recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1 (globin transcription factor 1); GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA1 GATA binding protein 1 (globin transcription factor 1) HGNC:4170 ERYF1 GF1 NFE1 GATA binding protein 1 GATA-binding protein 1 (globin transcription factor 1) erythrold transcription factor 1 globin transcription factor 1 transcription factor GATA1 GF-1 XLTT NF-E1 DNA-binding protein erythroid transcription factor 1; globin transcription factor 1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1
Gene Aliases: ERYF1; GATA-1; GATA1; GF-1; GF1; NF-E1; NFE1; XLANP; XLTDA; XLTT
UniProt ID: (Human) P15976, (Mouse) Q7TMX8
Entrez Gene ID: (Human) 2623, (Mouse) 14460
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.