Novus Biologicals
Manufacturer Code:NBP270799C
Catalog # NBP270799C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 300-449 of human GATA-6 (NP_005248) produced in E.coli. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GATA binding protein 6 GATA-binding factor 6 GATA-binding protein 6 transcription factor GATA-6; GATA-binding factor 6; Transcription factor GATA-6
Gene Aliases: GATA6
UniProt ID: (Human) Q92908
Entrez Gene ID: (Human) 2627
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.