Novus Biologicals
Manufacturer Code:NBP270797JF549
Catalog # N270797J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9F9 |
Immunogen | Recombinant protein fragment corresponding to amino acids 296-442 of human GATA-4 (NP_002043) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GATA binding protein 4 GATA-binding factor 4 GATA-binding protein 4 MGC126629 transcription factor GATA-4; GATA-binding factor 4; Transcription factor GATA-4
Gene Aliases: ASD2; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694
Entrez Gene ID: (Human) 2626
Molecular Function: nuclease nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.