Novus Biologicals
Manufacturer Code:NBP224585SS
Catalog # NBP224585SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 330-380 of human GATA-4 was used as the immunogen for the antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATA binding protein 4 GATA-binding factor 4 GATA-binding protein 4 MGC126629 transcription factor GATA-4; GATA-binding factor 4; Transcription factor GATA-4
Gene Aliases: ASD2; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694
Entrez Gene ID: (Human) 2626
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.