Novus Biologicals
Manufacturer Code:NBP147492
Catalog # NBP147492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F5 |
Immunogen | Purified recombinant full length human GATA1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Eryf1; Eryf1 ERYF1GATA-binding protein 1 (globin transcription factor 1) GATA binding protein 1 (globin transcription factor 1) GATA-1erythroid transcription factor GATA-binding factor 1 GF-1 GF1globin transcription factor 1 NF-E1 DNA-binding protein NFE1erythroid transcription factor 1 transcription factor GATA1 XLTT; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1 (globin transcription factor 1); GATA-1; GATA-binding factor 1; globin transcription factor 1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1
Gene Aliases: ERYF1; GATA-1; GATA1; GF-1; GF1; NF-E1; NFE1; XLANP; XLTDA; XLTT
UniProt ID: (Human) P15976
Entrez Gene ID: (Human) 2623
Molecular Function:
nuclease
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.