Novus Biologicals
Manufacturer Code:NBP226059
Catalog # NBP226059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-EQPQAGSSTSKE corresponding to C-Terminus according to NP_003033.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA transporter 1; GABA Transporter 1 GABATHG GABATRGAT-1 GABT1 GAT1sodium- and chloride-dependent GABA transporter 1 solute carrier family 6 (neurotransmitter transporter GABA) member 1 Solute carrier family 6 member 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: GABATHG; GABATR; GABT1; GAT1; MAE; SLC6A1
UniProt ID: (Human) P30531
Entrez Gene ID: (Human) 6529
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.