Novus Biologicals
Manufacturer Code:NBP159878
Catalog # NBP159878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC6A1(solute carrier family 6 (neurotransmitter transporter GABA) member 1) The peptide sequence was selected from the middle region of SLC6A1. Peptide sequence CCINSCTSMFAGFVIFSIVGFMAHVTKRSIADVAASGPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA transporter 1; GABA Transporter 1 GABATHG GABATRGAT-1 GABT1 GAT1sodium- and chloride-dependent GABA transporter 1 solute carrier family 6 (neurotransmitter transporter GABA) member 1 Solute carrier family 6 member 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: GABATHG; GABATR; GABT1; GAT1; MAE; SLC6A1
UniProt ID: (Human) P30531
Entrez Gene ID: (Human) 6529
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.