Invitrogen
Manufacturer Code:PA572882
Catalog # PIPA572882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GAS6 antibody for a 19 amino acid peptide from near the amino terminus of human GAS6. The immunogen is within amino acids 60 - 110 of GAS6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG AXSF AXL receptor tyrosine kinase ligand AXL stimulatory factor growth arrest-specific protein 6; GAS 6; GAS-6; GPF; growth arrest-specific 6; Growth arrest-specific protein 6; growth-potentiating factor
Gene Aliases: AXLLG; AXSF; Gas-6; GAS6
UniProt ID: (Human) Q14393, (Mouse) Q61592
Entrez Gene ID: (Human) 2621, (Rat) 58935, (Mouse) 14456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.