Abnova
Manufacturer Code:PAB17738
Catalog # 89-119-403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human GAS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GAS1.
For Research Use Only
Protein Aliases: GAS-1; growth arrest-specific 1; Growth arrest-specific gene-1; Growth arrest-specific protein 1
Gene Aliases: GAS1
UniProt ID: (Human) P54826
Entrez Gene ID: (Human) 2619
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.