Abnova
Manufacturer Code:MAB20074
Catalog # 89-336-474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | FFD-7 |
Immunogen | A synthetic peptide corresponding to human GART |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: 5'-phosphoribosylglycinamide transformylase; AIR synthase; AIRS; AIRS GARS GARTF MGC47764 PAIS PGFT PRGS; GAR transformylase; GARS; GART; glycinamide ribonucleotide formyltransferase; Glycinamide ribonucleotide synthetase; Phosphoribosyl-aminoimidazole synthetase; Phosphoribosylamine--glycine ligase; Phosphoribosylformylglycinamidine cyclo-ligase; Phosphoribosylglycinamide formyltransferase; Phosphoribosylglycinamide synthetase; Trifunctional purine biosynthetic protein adenosine-3
Gene Aliases: AIRS; GARS; GART; GARTF; PAIS; PGFT; PRGS
UniProt ID: (Human) P22102
Entrez Gene ID: (Human) 2618
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.