Abnova
Manufacturer Code:H00002618M01
Catalog # 89-022-269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D6-1D5 |
Immunogen | GART (AAH38958.1 1 a.a. ~ 433 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GART.
For Research Use Only
Protein Aliases: 5'-phosphoribosylglycinamide transformylase; AIR synthase; AIRS; GAR transformylase; GARS; GART; GARTF; glycinamide ribonucleotide formyltransferase; Glycinamide ribonucleotide synthetase; MGC47764; PAIS; PGFT; Phosphoribosyl-aminoimidazole synthetase; Phosphoribosylamine--glycine ligase; Phosphoribosylformylglycinamidine cyclo-ligase; Phosphoribosylglycinamide formyltransferase; Phosphoribosylglycinamide synthetase; PRGS; Trifunctional purine biosynthetic protein adenosine-3
Gene Aliases: AIRS; GARS; GART; GARTF; PAIS; PGFT; PRGS
UniProt ID: (Human) P22102
Entrez Gene ID: (Human) 2618
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.