Novus Biologicals
Manufacturer Code:NBP157774
Catalog # NBP157774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GART (phosphoribosylglycinamide formyltransferase phosphoribosylglycinamide synthetase phosphoribosylaminoimidazole synthetase) The peptide sequence was selected from the middle region of GART)(50ug). Immunogenizing p |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-phosphoribosylglycinamide transformylase; AIR synthase; AIRS; AIRS EC 6.3.3.1 EC 6.3.4.13 GARS GARTF MGC47764 PGFTEC 2.1.2.210PAIS phosphoribosylglycinamide formyltransferase phosphoribosylglycinamidesynthetase phosphoribosylaminoimidazole synthetase trifunctional purine biosynthetic protein adenosine-3; GAR transformylase; GARS; GART; glycinamide ribonucleotide formyltransferase; Glycinamide ribonucleotide synthetase; Phosphoribosyl-aminoimidazole synthetase; Phosphoribosylamine--glycine ligase; Phosphoribosylformylglycinamidine cyclo-ligase; Phosphoribosylglycinamide formyltransferase; Phosphoribosylglycinamide synthetase; Trifunctional purine biosynthetic protein adenosine-3
Gene Aliases: AIRS; GARS; GART; GARTF; PAIS; PGFT; PRGS
UniProt ID: (Human) P22102
Entrez Gene ID: (Human) 2618
Molecular Function:
hydrolase
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.