Invitrogen
Manufacturer Code:PA576739
Catalog # PIPA576739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GARS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-phosphoribosylglycinamide transformylaseAIR synthaseAIRSGAR transformylaseGARSGARTGARTFGlycinamide ribonucleotide synthetaseMGC47764PAISPGFTPhosphoribosyl-aminoimidazole synthetasePhosphoribosylglycinamide formyltransferasePhosphoribosylglycinamide formyltransferase phosphoribosylglycinamide synthetase phosphoribosylaminoimidazole synthetasePhosphoribosylglycinamide formyltransferase EC 2.1.2.29Phosphoribosylglycinamide formyltransferase phosphoribosylglycinamide synthetase phosphoribosylaminoimidazole synthetasePhosphoribosylglycinamide synthetasePRGSPUR2&lowbarHUMANTrifunctional purine biosynthetic protein adenosine 3; AP-4-A synthetase; Ap4A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; Diadenosine tetraphosphate synthetase; glycine tRNA ligase; Glycine--tRNA ligase; Glycyl-tRNA synthetase; Glycyl-tRNA synthetase 1; GlyRS; storage granule protein 23
Gene Aliases: CMT2D; DSMAV; GARS; GARS1; Gena201; GENA202; GlyRS; HMN5; Nmf249; RGD1559871; Sgrp23; SMAD1
UniProt ID: (Human) P41250, (Rat) Q5I0G4, (Mouse) Q9CZD3
Entrez Gene ID: (Human) 2617, (Rat) 297113, (Mouse) 353172
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.