Invitrogen
Manufacturer Code:PA5106927
Catalog # PIPA5106927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GARS corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6.1.1.14 AIR synthase AIRS Ap4A synthetase AP-4-A synthetase Charcot-Marie-Tooth neuropathy 2D Charcot-Marie-Tooth neuropathy neuronal type D CMT2D Diadenosine tetraphosphate synthetase DSMAV EC 2.1.2.29 GAR transformylase Gars GART GARTF Gena201 GENA202 glycine tRNA ligase glycine--tRNA ligase Glycyl-tRNA synthetase glyRS HMN5 MGC47764 Nmf249 PAIS PGFT PRGS RGD1559871 Sgrp23 SMAD1 storage granule protein 23; AP-4-A synthetase; Ap4A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; Diadenosine tetraphosphate synthetase; glycine tRNA ligase; Glycine--tRNA ligase; Glycyl-tRNA synthetase; Glycyl-tRNA synthetase 1; GlyRS; storage granule protein 23
Gene Aliases: CMT2D; DSMAV; GARS; GARS1; Gena201; GENA202; GlyRS; HMN5; Nmf249; RGD1559871; Sgrp23; SMAD1
UniProt ID: (Human) P41250, (Rat) Q5I0G4, (Mouse) Q9CZD3
Entrez Gene ID: (Human) 2617, (Rat) 297113, (Mouse) 353172
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.