Proteintech
Manufacturer Code:117111AP150UL
Catalog # 50-172-7394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GAR1 Fusion Protein Ag2282 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAR1 homolog, ribonucleoprotein; GAR1 NOLA1 snoRNP protein GAR1; GAR1 ribonucleoprotein homolog; H/ACA ribonucleoprotein complex subunit 1; H/ACA small nucleolar RNPs; Nucleolar protein family A member 1; nucleolar protein family A, member 1 (H/ACA small nucleolar RNPs); snoRNP protein GAR1
Gene Aliases: AA409823; AI326794; C430047J18Rik; GAR1; NOLA1
UniProt ID: (Human) Q9NY12, (Mouse) Q9CY66, (Rat) Q6AYA1
Entrez Gene ID: (Human) 54433, (Mouse) 68147, (Rat) 499709
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.