Invitrogen
Manufacturer Code:MA515738A488
Catalog # MA515738A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GA1R |
Immunogen | recombit GAPDH |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; EC 1.2.1.12 G3PD G3PDH GAPD glyceraldehyde-3-phosphate dehydrogenase HGNC:4141 MGC88685 GAPDH Glyceraldehyde-3-phosphate dehydrogenase; epididymis secretory sperm binding protein Li 162eP; GAPDH; glceraldehyde-3-phosphate dehydrogenase; Glyceraldehyde-3-phosphate dehydrogenase; multifunctional protein, glycolytic enzyme; Peptidyl-cysteine S-nitrosylase GAPDH; unnamed protein product
Gene Aliases: BARS-38; CDABP0047; G3PD; GAPD; GAPDH; HEL-S-162eP; I79_001391; KNC-NDS6; OK/SW-cl.12
UniProt ID: (Rabbit) P46406, (Human) P04406, (Chicken) P00356, (Mouse) P16858, (Chinese hamster) P17244, (Rat) P04797
Entrez Gene ID: (Rabbit) 100009074, (Human) 2597, (Chicken) 374193, (Mouse) 14433, (Chinese hamster) 100736557, (Rat) 24383
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.