OriGene
Manufacturer Code:CF802563
Catalog # 50-166-6053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F8 |
Immunogen | Full length human recombit protein of human GAPDH produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aging-associated gene 9 protein; epididymis secretory sperm binding protein Li 162eP; G3PD GAPD HEL-S-162eP; GAPDH; Glyceraldehyde-3-phosphate dehydrogenase; Peptidyl-cysteine S-nitrosylase GAPDH
Gene Aliases: CDABP0047; G3PD; GAPD; GAPDH; HEL-S-162eP; OK/SW-cl.12
UniProt ID: (Human) P04406
Entrez Gene ID: (Human) 2597
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.