Invitrogen
Manufacturer Code:MA116757
Catalog # AMPA6757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D4 |
Immunogen | Extensively purified pig GAPDH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; epididymis secretory sperm binding protein Li 162eP; GAPD; GAPDH; glyceraldehyde phosphate dehydrogenase; Glyceraldehyde-3-phosphate dehydrogenase; Glyceraldehyde-3-Phosphate Dehydrogenase GAPD G3PD G3PDH; Peptidyl-cysteine S-nitrosylase GAPDH
Gene Aliases: BARS-38; bb02e05; cb609; CDABP0047; G3PD; G3PDH; GAPD; GAPDH; HEL-S-162eP; I79_001391; mg:bb02e05; OK/SW-cl.12; wu:fb33a10; wu:ft80f05
UniProt ID: (Human) P04406, (Bovine) P10096, (Pig) P00355, (Mouse) P16858, (Chinese hamster) P17244, (Rat) P04797, (Zebrafish) Q5XJ10
Entrez Gene ID: (Sheep) 443005, (Human) 2597, (Bovine) 281181, (Pig) 396823, (Mouse) 14433, (Chinese hamster) 100736557, (Rat) 24383, (Zebrafish) 317743
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.