Novus Biologicals
Manufacturer Code:NB3003270.025ML
Catalog # NB018158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This GAPDH antibody was developed against full length recombinant human GAPDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aging-associated gene 9 protein; aging-associated gene 9 protein EC 1.2.1 EC 1.2.1.12 EC 2.6.99.- G3PD GAPD glyceraldehyde 3-phosphate dehydrogenase glyceraldehyde-3-phosphate dehydrogenase MGC88685 Peptidyl-cysteine S-nitrosylase GAPDH; epididymis secretory sperm binding protein Li 162eP; GAPDH; Glyceraldehyde-3-phosphate dehydrogenase; Peptidyl-cysteine S-nitrosylase GAPDH
Gene Aliases: CDABP0047; G3PD; GAPD; GAPDH; HEL-S-162eP; OK/SW-cl.12
UniProt ID: (Human) P04406
Entrez Gene ID: (Human) 2597
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.