Invitrogen
Manufacturer Code:PA522899
Catalog # PIPA522899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 216-226 of rat/mouse GAP-43 (C)-KEDPEADQEHA coupled to maleimide-activated KLH. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axonal membrane protein GAP-43; B-50 PP46 axonal membrane protein GAP-43 calmodulin-binding protein P-57 growth-associated protein 43 nerve growth-related peptide GAP43 neural phosphoprotein B-50 neuromodulin neuron growth-associated protein 43 protein F1; brain abundant, membrane attached signal protein 2; Calmodulin-binding protein P-57; growth accentuating protein 43; Growth-associated protein 43; nerve growth-related peptide GAP43; Neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; pp46; Protein F1
Gene Aliases: B-50; Basp2; GAP-43; GAP43; PP46
UniProt ID: (Human) P17677, (Rat) P07936, (Mouse) P06837
Entrez Gene ID: (Chicken) 427955, (Dog) 478572, (Human) 2596, (Pig) 100623423, (Rat) 29423, (Mouse) 14432
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.