Novus Biologicals
Manufacturer Code:NB300143V
Catalog # NB300143V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminal peptide of rodent GAP43 KEDPEADQEHA coupled to KLH. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axonal membrane protein GAP-43; Axonal membrane protein GAP-43 B-50 calmodulin-binding protein P-57 growth associated protein 43 Growth-associated protein 43 nerve growth-related peptide GAP43 Neural phosphoprotein B-50 neuromodulin neuron growth-associated protein 43 PP46 protein F1; calmodulin-binding protein P-57; Growth-associated protein 43; nerve growth-related peptide GAP43; Neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; pp46; protein F1
Gene Aliases: B-50; GAP43; PP46
UniProt ID: (Human) P17677
Entrez Gene ID: (Human) 2596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.