Invitrogen
Manufacturer Code:PA599628
Catalog # PIPA599628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Glucosidase II alpha corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha glucosidase 2 alpha neutral subunit alpha-glucosidase 2 AU042638 carbohydrate processing enzyme of the endoplasmic reticulum G2AN GANAB GIIA glucosidase II alpha subunit glucosidase II alpha-subunit glucosidase II subunit alpha glucosidase alpha neutral AB GluII Kiaa0088 mKIAA0088 neutral AB neutral alpha-glucosidase AB; Alpha-glucosidase 2; carbohydrate processing enzyme of the endoplasmic reticulum; glucosidase II alpha-subunit; Glucosidase II subunit alpha; Neutral alpha-glucosidase AB
Gene Aliases: AU042638; G2AN; GANAB; GIIA; GLUII; KIAA0088; mKIAA0088
UniProt ID: (Human) Q14697, (Mouse) Q8BHN3
Entrez Gene ID: (Human) 23193, (Mouse) 14376
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.