Invitrogen
Manufacturer Code:PA521425
Catalog # PIPA521425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 654 and 944 of alpha Glucosidase II (Uniprot IDNo.Q14697) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-glucosidase 2; G2AN GLUII alpha glucosidase II alpha subunit alpha-glucosidase 2 glucosidase II subunit alpha neutral alpha-glucosidase AB; Glucosidase II subunit alpha; Neutral alpha-glucosidase AB
Gene Aliases: G2AN; GANAB; GIIA; GLUII; KIAA0088
UniProt ID: (Human) Q14697
Entrez Gene ID: (Human) 23193
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.