Proteintech
Manufacturer Code:663221IG150UL
Catalog # 50-173-6754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E7B11 |
Immunogen | GAMT Fusion Protein Ag16778 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 20; GAMT PIG2 TP53I2; guanidinoacetate methyltransferase; Guanidinoacetate N-methyltransferase
Gene Aliases: AA571402; CCDS2; GAMT; GMT; HEL-S-20; PIG2; Spintz1; TP53I2
UniProt ID: (Human) Q14353, (Mouse) O35969
Entrez Gene ID: (Human) 2593, (Pig) 100625146, (Mouse) 14431, (Rat) 25257
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.