Invitrogen
Manufacturer Code:PA5100538
Catalog # PIPA5100538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GAMT corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA571402 CCDS2 epididymis secretory protein Li 20 GAMT GMT guanidinoacetate methyltransferase guanidinoacetate N-methyltransferase HEL-S-20 PIG2 Spintz1 TP53I2 zgc:123136 zgc:123136 protein; epididymis secretory protein Li 20; GAMT; guanidinoacetate methyltransferase; Guanidinoacetate N-methyltransferase
Gene Aliases: AA571402; CCDS2; GAMT; GMT; HEL-S-20; PIG2; Spintz1; TP53I2; zgc:123136
UniProt ID: (Human) Q14353, (Mouse) O35969, (Zebrafish) Q71N41
Entrez Gene ID: (Human) 2593, (Mouse) 14431, (Rat) 25257, (Zebrafish) 796865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.